Assay Details
Gene Symbol: Shf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008762216.1 | XP_008760438.1 | 1 - 2 | 285 | 108 | |
XM_008762218.1 | XP_008760440.1 | 1 - 2 | 285 | 108 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008762216.1, XM_008762218.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
