Assay Details
Gene Symbol: Shf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234884.2 | XP_006234946.1 | 3 - 4 | 507 | 62 | |
XM_008762216.1 | XP_008760438.1 | 5 - 6 | 951 | 62 | ||
XM_008762218.1 | XP_008760440.1 | 5 - 6 | 951 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
