Assay Details
Gene Symbol: Pacsin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009966.2 | NP_001009966.1 | 4 - 5 | 541 | 78 | |
XM_006234509.2 | XP_006234571.1 | 5 - 6 | 740 | 78 | ||
XM_006234510.2 | XP_006234572.1 | 5 - 6 | 661 | 78 | ||
XM_006234511.2 | XP_006234573.1 | 4 - 5 | 699 | 78 | ||
XM_008762007.1 | XP_008760229.1 | 5 - 6 | 727 | 78 | ||
GenBank mRNA | AY858802.1 | - | 3 - 4 | 451 | 78 | |
BC100636.1 | - | 4 - 5 | 541 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
