Assay Details
Gene Symbol: Pacsin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009966.2 | NP_001009966.1 | 8 - 9 | 1122 | 128 | |
XM_006234509.2 | XP_006234571.1 | 9 - 10 | 1321 | 128 | ||
XM_006234510.2 | XP_006234572.1 | 9 - 10 | 1242 | 128 | ||
XM_006234511.2 | XP_006234573.1 | 8 - 9 | 1280 | 128 | ||
XM_008762007.1 | XP_008760229.1 | 9 - 10 | 1308 | 128 | ||
GenBank mRNA | AY858802.1 | - | 7 - 8 | 1032 | 128 | |
BC100636.1 | - | 8 - 9 | 1122 | 128 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
