Assay Details
Gene Symbol: Adal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008762166.1 | XP_008760388.1 | 4 - 5 | 547 | 92 | |
XM_008762169.1 | XP_008760391.1 | 3 - 4 | 283 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
