Assay Details
Gene Symbol: Adal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014047.1 | NP_001014069.1 | 1 - 2 | 274 | 63 | |
XM_008762166.1 | XP_008760388.1 | 6 - 7 | 752 | 63 | ||
XM_008762167.1 | XP_008760389.1 | 4 - 5 | 282 | 63 | ||
XM_008762168.1 | XP_008760390.1 | 2 - 3 | 183 | 63 | ||
XM_008762169.1 | XP_008760391.1 | 5 - 6 | 488 | 63 | ||
GenBank mRNA | BC089959.1 | - | 1 - 2 | 274 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
