Assay Details
Gene Symbol: Nr1h3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031627.2 | NP_113815.2 | 7 - 8 | 1115 | 76 | |
XM_006234533.2 | XP_006234595.1 | 8 - 9 | 1293 | 76 | ||
XM_006234534.1 | XP_006234596.1 | 8 - 9 | 1234 | 76 | ||
XM_006234535.2 | XP_006234597.1 | 8 - 9 | 1435 | 76 | ||
XM_006234537.1 | XP_006234599.1 | 7 - 8 | 905 | 76 | ||
XM_008762013.1 | XP_008760235.1 | 7 - 8 | 1025 | 76 | ||
GenBank mRNA | BC088750.1 | - | 7 - 8 | 1115 | 76 | |
U11685.1 | - | 7 - 8 | 1122 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
