Assay Details
Gene Symbol: Pla2g4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001080051.5 | XP_001080051.2 | 17 - 18 | 1949 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
