Assay Details
Gene Symbol: Casc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170594.1 | NP_001164065.1 | 15 - 16 | 5585 | 117 | |
XM_008762095.1 | XP_008760317.1 | 18 - 19 | 5771 | 117 | ||
XM_008762096.1 | XP_008760318.1 | 18 - 19 | 6001 | 117 | ||
XM_008762097.1 | XP_008760319.1 | 18 - 19 | 5814 | 117 | ||
XM_008762098.1 | XP_008760320.1 | 16 - 17 | 5684 | 117 | ||
XM_008762099.1 | XP_008760321.1 | 18 - 19 | 5758 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
