Assay Details
Gene Symbol: Casc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170594.1 | NP_001164065.1 | 16 - 17 | 5621 | 70 | |
XM_008762095.1 | XP_008760317.1 | 19 - 20 | 5807 | 70 | ||
XM_008762096.1 | XP_008760318.1 | 19 - 20 | 6037 | 70 | ||
XM_008762097.1 | XP_008760319.1 | 19 - 20 | 5850 | 70 | ||
XM_008762098.1 | XP_008760320.1 | 17 - 18 | 5720 | 70 | ||
XM_008762099.1 | XP_008760321.1 | 19 - 20 | 5794 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
