Assay Details
Gene Symbol: Acp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016988.2 | NP_058684.2 | 4 - 5 | 490 | 58 | |
XM_006234500.1 | XP_006234562.1 | 5 - 6 | 1412 | 58 | ||
GenBank mRNA | BC081823.1 | - | 4 - 5 | 461 | 58 | |
CB720349.1 | - | 4 - 5 | 490 | 58 | ||
M27893.1 | - | 4 - 5 | 459 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
