Assay Details
Gene Symbol: Disp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107759.2 | NP_001101229.2 | 6 - 7 | 952 | 71 | |
XM_006234756.2 | XP_006234818.1 | 6 - 7 | 966 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
