Assay Details
Gene Symbol: Psmc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031595.1 | NP_113783.1 | 10 - 11 | 1360 | 70 | |
GenBank mRNA | BC062019.1 | - | 10 - 11 | 1270 | 70 | |
D83522.1 | - | 10 - 11 | 1234 | 70 | ||
FQ225567.1 | - | 9 - 10 | 1019 | 70 | ||
FQ225947.1 | - | 9 - 10 | 1019 | 70 | ||
FQ226273.1 | - | 9 - 10 | 1019 | 70 | ||
FQ232987.1 | - | 9 - 10 | 1020 | 70 | ||
U77918.1 | - | 10 - 11 | 1360 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
