Assay Details
Gene Symbol: Il1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017019.1 | NP_058715.1 | 1 - 2 | 52 | 171 | |
GenBank mRNA | D00403.1 | - | 1 - 2 | 52 | 171 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the D00403.1,NM_017019.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
