Assay Details
Gene Symbol: Duoxa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107767.1 | NP_001101237.1 | 6 - 7 | 922 | 84 | |
XM_006234857.1 | XP_006234919.1 | 7 - 8 | 810 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
