Assay Details
Gene Symbol: Galnt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015032.3 | NP_001015032.2 | 1 - 2 | 622 | 67 | |
XM_006234290.2 | XP_006234352.1 | 2 - 3 | 857 | 67 | ||
GenBank mRNA | BC101886.1 | - | 1 - 2 | 625 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
