Assay Details
Gene Symbol: Galnt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015032.3 | NP_001015032.2 | 3 - 4 | 949 | 68 | |
XM_006234290.2 | XP_006234352.1 | 4 - 5 | 1184 | 68 | ||
GenBank mRNA | AB040674.1 | - | 3 - 4 | 838 | 68 | |
BC101886.1 | - | 3 - 4 | 952 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
