Assay Details
Gene Symbol: Galnt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015032.3 | NP_001015032.2 | 5 - 6 | 1307 | 148 | |
XM_006234290.2 | XP_006234352.1 | 6 - 7 | 1542 | 148 | ||
GenBank mRNA | AB040674.1 | - | 5 - 6 | 1196 | 148 | |
BC101886.1 | - | 5 - 6 | 1310 | 148 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
