Assay Details
Gene Symbol: Galnt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015032.3 | NP_001015032.2 | 8 - 9 | 1734 | 100 | |
XM_006234290.2 | XP_006234352.1 | 9 - 10 | 1969 | 100 | ||
GenBank mRNA | AB040674.1 | - | 8 - 9 | 1623 | 100 | |
BC101886.1 | - | 8 - 9 | 1737 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
