Assay Details
Gene Symbol: Ppt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022502.2 | NP_071947.1 | 7 - 8 | 726 | 101 | |
GenBank mRNA | BC126089.1 | - | 7 - 8 | 726 | 101 | |
FQ213911.1 | - | 738 | 101 | |||
L34262.1 | - | 724 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
