Assay Details
Gene Symbol: Ttll10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024758.2 | NP_001019929.1 | 5 - 6 | 495 | 79 | |
NM_001170533.1 | NP_001164004.1 | 6 - 7 | 917 | 79 | ||
XM_006239559.2 | XP_006239621.1 | 6 - 7 | 1077 | 79 | ||
XM_006239562.2 | XP_006239624.1 | 4 - 5 | 725 | 79 | ||
XM_008764356.1 | XP_008762578.1 | 7 - 8 | 2364 | 79 | ||
XM_008764357.1 | XP_008762579.1 | 6 - 7 | 873 | 79 | ||
GenBank mRNA | BC083833.1 | - | 3 - 4 | 334 | 79 | |
BC097417.1 | - | 5 - 6 | 495 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
