Assay Details
Gene Symbol: Sdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100539.1 | NP_001094009.1 | 2 - 3 | 229 | 103 | |
GenBank mRNA | BC158620.1 | - | 2 - 3 | 229 | 103 | |
DQ403001.1 | - | 1 - 2 | 134 | 103 | ||
FQ215046.1 | - | 2 - 3 | 225 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
