Assay Details
Gene Symbol: Sdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100539.1 | NP_001094009.1 | 3 - 4 | 314 | 71 | |
GenBank mRNA | BC158620.1 | - | 3 - 4 | 314 | 7446362 | 71 |
DQ403001.1 | - | 2 - 3 | 219 | 71 | ||
FQ215046.1 | - | 3 - 4 | 310 | 71 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
