Assay Details
Gene Symbol: Sdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100539.1 | NP_001094009.1 | 4 - 5 | 454 | 94 | |
GenBank mRNA | BC158620.1 | - | 4 - 5 | 454 | 94 | |
DQ403001.1 | - | 3 - 4 | 359 | 94 | ||
FQ215046.1 | - | 4 - 5 | 450 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
