Assay Details
Gene Symbol: Sdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100539.1 | NP_001094009.1 | 5 - 6 | 568 | 69 | |
GenBank mRNA | BC158620.1 | - | 5 - 6 | 568 | 69 | |
DQ403001.1 | - | 4 - 5 | 473 | 69 | ||
FQ215046.1 | - | 5 - 6 | 564 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
