Assay Details
Gene Symbol: Rhbdl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106684.1 | NP_001100154.1 | 1 - 2 | 145 | 82 | |
XM_006238802.2 | XP_006238864.1 | 3 - 4 | 753 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001106684.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
