Assay Details
Gene Symbol: Rhbdl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106684.1 | NP_001100154.1 | 2 - 3 | 262 | 121 | |
XM_006238802.2 | XP_006238864.1 | 4 - 5 | 870 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
