Assay Details
Gene Symbol: Rhbdl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106684.1 | NP_001100154.1 | 5 - 6 | 490 | 82 | |
XM_006238802.2 | XP_006238864.1 | 7 - 8 | 1098 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
