Assay Details
Gene Symbol: Stk40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183056.2 | NP_898879.2 | 5 - 6 | 731 | 91 | |
XM_006238881.2 | XP_006238943.1 | 5 - 6 | 2441 | 91 | ||
GenBank mRNA | AY336055.1 | - | 5 - 6 | 733 | 91 | |
BC100062.1 | - | 5 - 6 | 731 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
