Assay Details
Gene Symbol: Stk40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183056.2 | NP_898879.2 | 6 - 7 | 782 | 76 | |
XM_006238881.2 | XP_006238943.1 | 6 - 7 | 2492 | 76 | ||
GenBank mRNA | AY336055.1 | - | 6 - 7 | 784 | 76 | |
BC100062.1 | - | 6 - 7 | 782 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
