Assay Details
Gene Symbol: Srsf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025738.1 | NP_001020909.1 | 4 - 5 | 537 | 107 | |
XM_006239220.2 | XP_006239282.1 | 4 - 5 | 580 | 107 | ||
XM_006239222.2 | XP_006239284.1 | 4 - 5 | 578 | 107 | ||
GenBank mRNA | BC098831.1 | - | 4 - 5 | 537 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
