Assay Details
Gene Symbol: Srsf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025738.1 | NP_001020909.1 | 1 - 2 | 162 | 152 | |
XM_006239220.2 | XP_006239282.1 | 1 - 2 | 205 | 152 | ||
XM_006239221.2 | XP_006239283.1 | 1 - 2 | 205 | 152 | ||
XM_006239222.2 | XP_006239284.1 | 1 - 2 | 203 | 152 | ||
XM_006239223.2 | XP_006239285.1 | 1 - 2 | 204 | 152 | ||
GenBank mRNA | BC098831.1 | - | 1 - 2 | 162 | 152 | |
FQ227288.1 | - | 1 - 2 | 183 | 152 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC098831.1, NM_001025738.1, XM_006239220.2, XM_006239221.2, XM_006239222.2, XM_006239223.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
