Assay Details
Gene Symbol: Cdk11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145766.2 | NP_665709.2 | 19 - 20 | 2309 | 56 | |
XM_006239527.2 | XP_006239589.1 | 18 - 19 | 2865 | 56 | ||
XM_006239528.2 | XP_006239590.1 | 19 - 20 | 2672 | 56 | ||
XM_006239529.2 | XP_006239591.1 | 19 - 20 | 2593 | 56 | ||
XM_006239530.1 | XP_006239592.1 | 19 - 20 | 2293 | 56 | ||
XM_006239531.2 | XP_006239593.1 | 19 - 20 | 2397 | 56 | ||
XM_006239532.2 | XP_006239594.1 | 14 - 15 | 2741 | 56 | ||
XM_006239533.1 | XP_006239595.1 | 14 - 15 | 1718 | 56 | ||
GenBank mRNA | AA817907.1 | - | 1 - 2 | 56 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
