Assay Details
Gene Symbol: Ppie | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047868.1 | NP_001041333.1 | 3 - 4 | 169 | 98 | |
XM_006238796.2 | XP_006238858.1 | 2 - 3 | 184 | 98 | ||
XM_006238797.2 | XP_006238859.1 | 2 - 3 | 193 | 98 | ||
GenBank mRNA | AY325152.1 | - | 3 - 4 | 169 | 98 | |
BC158560.1 | - | 2 - 3 | 134 | 9102303 | 98 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the XM_006238797.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
