Assay Details
Gene Symbol: Ppie | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047868.1 | NP_001041333.1 | 6 - 7 | 323 | 94 | |
XM_006238796.2 | XP_006238858.1 | 5 - 6 | 338 | 94 | ||
XM_006238797.2 | XP_006238859.1 | 5 - 6 | 347 | 94 | ||
GenBank mRNA | AY325152.1 | - | 6 - 7 | 323 | 94 | |
BC158560.1 | - | 5 - 6 | 288 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
