Assay Details
Gene Symbol: Ppie | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238796.2 | XP_006238858.1 | 9 - 10 | 893 | 60 | |
XM_006238797.2 | XP_006238859.1 | 9 - 10 | 902 | 60 | ||
GenBank mRNA | BC158560.1 | - | 9 - 10 | 843 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
