Assay Details
Gene Symbol: Hivep3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107972.1 | NP_001101442.1 | 1 - 2 | 131 | 69 | |
XM_006238779.2 | XP_006238841.1 | 2 - 3 | 3283 | 69 | ||
XM_008764014.1 | XP_008762236.1 | 1 - 2 | 3380 | 69 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001107972.1,XM_006238779.2,XM_008764014.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
