Assay Details
Gene Symbol: Hivep3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107972.1 | NP_001101442.1 | 4 - 5 | 5928 | 65 | |
XM_006238779.2 | XP_006238841.1 | 5 - 6 | 9080 | 65 | ||
XM_008764014.1 | XP_008762236.1 | 4 - 5 | 9177 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
