Assay Details
Gene Symbol: Smap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100669.1 | NP_001094139.1 | 8 - 9 | 1213 | 68 | |
GenBank mRNA | BC099210.1 | - | 1 - 2 | 76 | 68 | |
BC161927.1 | - | 8 - 9 | 1207 | 8374170 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
