Assay Details
Gene Symbol: Smap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100669.1 | NP_001094139.1 | 9 - 10 | 1529 | 70 | |
GenBank mRNA | BC099210.1 | - | 2 - 3 | 392 | 70 | |
BC161927.1 | - | 9 - 10 | 1523 | 8374170 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P142549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937220 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735071 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633461 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958218 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248391 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453546 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 98643 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135460 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313086 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549875 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445746 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111192 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789059 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68342 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38039 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86912 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317101 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225984 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876713 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349635 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101585 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766314 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500793 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273050 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311808 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483033 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500265 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610919 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422334 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272371 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955920 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488449 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341107 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861216 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792935 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27465 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81771 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139154 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 45548 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P027268 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P027268 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P027268 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P516003 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 8905 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10879459 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10879454 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10879461 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10879456 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10879455 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10879458 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10879457 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10879463 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10879462 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10879460 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10879453 | - | - |
Rat Genome 230 2.0 Array | 1398062_at | - | - |
Rat Genome 230 2.0 Array | 1372561_at | - | - |
Rat Genome 230 2.0 Array | 1391204_PM_at | - | - |
Rat Genome 230 2.0 Array | 1390350_at | - | - |
Rat Genome 230 2.0 Array | 1391204_at | - | - |
Rat Genome 230 2.0 Array | 1372561_PM_at | - | - |
Rat Genome 230 2.0 Array | 1398062_PM_at | - | - |
Rat Genome 230 2.0 Array | 1390350_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1372561_at | - | - |
Affymetrix HT Rat Focus Array | 1398062_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1390350_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1372561_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1390350_at | - | - |
Affymetrix HT Rat Focus Array | 1391204_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1398062_at | - | - |
Affymetrix HT Rat Focus Array | 1391204_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0500026127.rn.1 | TC0500003530.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500026128.rn.1 | TC0500003530.rn.1 | RTA TC |
Rat Expression Set 230 | 1390350_PM_at | - | - |
Rat Expression Set 230 | 1372561_PM_at | - | - |
Rat Expression Set 230 | 1372561_at | - | - |
Rat Expression Set 230 | 1390350_at | - | - |
Rat Expression Set 230 | 1398062_PM_at | - | - |
Rat Expression Set 230 | 1398062_at | - | - |
Rat Expression Set 230 | 1391204_at | - | - |
Rat Expression Set 230 | 1391204_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10879453 | - | - |
Rat Genome U34 Set | rc_AI234095_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21468204 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1350057 | - | - |
