Assay Details
Gene Symbol: Acot7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146061.1 | NP_001139533.1 | 3 - 4 | 500 | 65 | |
NM_013214.4 | NP_037346.2 | 3 - 4 | 428 | 65 | ||
XM_006239447.2 | XP_006239509.1 | 3 - 4 | 528 | 65 | ||
XM_006239448.1 | XP_006239510.1 | 3 - 4 | 436 | 65 | ||
GenBank mRNA | BC087716.1 | - | 3 - 4 | 380 | 7316866 | 65 |
D88891.1 | - | 3 - 4 | 500 | 65 | ||
Y09332.1 | - | 3 - 4 | 328 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
