Assay Details
Gene Symbol: Zmym1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107983.1 | NP_001101453.1 | 5 - 6 | 864 | 95 | |
XM_006238878.2 | XP_006238940.1 | 3 - 4 | 2068 | 95 | ||
XM_006238879.1 | XP_006238941.1 | 5 - 6 | 778 | 95 | ||
XM_006238880.2 | XP_006238942.1 | 4 - 5 | 802 | 95 | ||
GenBank mRNA | BC160845.1 | - | 4 - 5 | 757 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
