Assay Details
Gene Symbol: Lck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100709.1 | NP_001094179.1 | 2 - 3 | 192 | 100 | |
XM_006238932.2 | XP_006238994.1 | 3 - 4 | 359 | 100 | ||
XM_006238933.2 | XP_006238995.1 | 3 - 4 | 321 | 100 | ||
XM_008764140.1 | XP_008762362.1 | 3 - 4 | 377 | 100 | ||
GenBank mRNA | BC160881.1 | - | 2 - 3 | 192 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
