Assay Details
Gene Symbol: Agmat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048185.1 | NP_001041650.1 | 6 - 7 | 1005 | 70 | |
GenBank mRNA | BC091231.1 | - | 6 - 7 | 989 | 70 | |
BC105628.1 | - | 6 - 7 | 1005 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
