Assay Details
Gene Symbol: Mxra8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007002.1 | NP_001007003.1 | 4 - 5 | 508 | 92 | |
XM_006239574.1 | XP_006239636.1 | 4 - 5 | 876 | 92 | ||
XM_006239575.1 | XP_006239637.1 | 4 - 5 | 867 | 92 | ||
GenBank mRNA | BC083849.1 | - | 4 - 5 | 508 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
