Assay Details
Gene Symbol: Cdc42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008764287.1 | XP_008762509.1 | 1 - 2 | 103 | 91 | |
GenBank mRNA | FQ225541.1 | - | 1 - 2 | 135 | 91 | |
FQ225973.1 | - | 1 - 2 | 135 | 91 | ||
FQ228149.1 | - | 1 - 2 | 135 | 91 | ||
FQ231846.1 | - | 1 - 2 | 135 | 91 | ||
FQ233586.1 | - | 1 - 2 | 33 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008764287.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
