Assay Details
Gene Symbol: Igsf21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271454.1 | NP_001258383.1 | 4 - 5 | 992 | 67 | |
XM_008764231.1 | XP_008762453.1 | 3 - 4 | 306 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
