Assay Details
Gene Symbol: Igsf21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271454.1 | NP_001258383.1 | 5 - 6 | 1468 | 68 | |
XM_008764231.1 | XP_008762453.1 | 4 - 5 | 782 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
