Assay Details
Gene Symbol: Fam76a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108686.1 | NP_001102156.1 | 5 - 6 | 696 | 140 | |
GenBank mRNA | FQ222884.1 | - | 5 - 6 | 698 | 140 |

More Information
Set Membership: |
![]() ![]() ![]() |
