Assay Details
Gene Symbol: Ttll10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024758.2 | NP_001019929.1 | 12 - 13 | 1403 | 82 | |
NM_001170533.1 | NP_001164004.1 | 13 - 14 | 1825 | 82 | ||
XM_006239559.2 | XP_006239621.1 | 13 - 14 | 1985 | 82 | ||
XM_006239562.2 | XP_006239624.1 | 11 - 12 | 1633 | 82 | ||
XM_008764356.1 | XP_008762578.1 | 14 - 15 | 3272 | 82 | ||
XM_008764357.1 | XP_008762579.1 | 13 - 14 | 1781 | 82 | ||
GenBank mRNA | BC083833.1 | - | 10 - 11 | 1242 | 82 | |
BC097417.1 | - | 12 - 13 | 1403 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
